siRNA Details
| Gene Symbol: Brd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012047.2 | NP_036177.1 | 2, 3 | 496 | ||
| GenBank mRNA | AF084259.1 | - | 2, 3 | 469 | ||
| AK138934.1 | - | 2, 3 | 500 | |||
| AK142758.1 | - | 2, 3 | 482 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Brd7| Gene Symbol: Brd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012047.2 | NP_036177.1 | 2, 3 | 496 | ||
| GenBank mRNA | AF084259.1 | - | 2, 3 | 469 | ||
| AK138934.1 | - | 2, 3 | 500 | |||
| AK142758.1 | - | 2, 3 | 482 | |||