siRNA Details
| Gene Symbol: Sgsh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_018822.3 | NP_061292.2 | 4 | 483 | ||
| GenBank mRNA | AF153827.1 | - | 4 | 457 | ||
| AF304053.1 | - | 4 | 457 | |||
| AF304054.1 | - | 4 | 457 | |||
| AK034220.1 | - | 4 | 394 | |||
| AK052266.1 | - | 4 | 476 | |||
| AK128926.1 | - | 4 | 483 | |||
| AK141266.1 | - | 4 | 431 | |||
| BC130268.1 | - | 4 | 480 | 40143074 | ||

Sgsh
Genomic Map