siRNA Details
| Gene Symbol: G3bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013716.2 | NP_038744.1 | 6 | 587 | ||
| GenBank mRNA | AB001927.1 | - | 6 | 587 | ||
| AK141689.1 | - | 6 | 654 | |||
| AK208718.1 | - | 3 | 204 | |||
| AK220230.1 | - | 6 | 641 | |||
| BC021156.1 | - | 6 | 599 | 4020362 | ||

G3bp1
Genomic Map