siRNA Details
| Gene Symbol: Aoah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012054.4 | NP_036184.1 | 15, 16 | 1405 | ||
| GenBank mRNA | AK084452.1 | - | 15, 16 | 1400 | ||
| AK085343.1 | - | 15, 16 | 1377 | |||
| AK133301.1 | - | 15, 16 | 1346 | |||
| AK170293.1 | - | 14, 15 | 941 | |||
| AK171572.1 | - | 15, 16 | 1224 | |||
| CT010242.1 | - | 15, 16 | 1124 | |||

Aoah
Genomic Map