siRNA Details
| Gene Symbol: Zfp275 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160229.1 | NP_001153701.1 | 5 | 631 | ||
| NM_031494.2 | NP_113682.2 | 6 | 724 | |||
| GenBank mRNA | AK142957.1 | - | 5 | 631 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp275| Gene Symbol: Zfp275 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160229.1 | NP_001153701.1 | 5 | 631 | ||
| NM_031494.2 | NP_113682.2 | 6 | 724 | |||
| GenBank mRNA | AK142957.1 | - | 5 | 631 | ||