siRNA Details
| Gene Symbol: Eif2ak4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177806.1 | NP_001171277.1 | 20, 21 | 2953 | ||
| NM_013719.3 | NP_038747.2 | 21, 22 | 3257 | |||
| GenBank mRNA | AJ243533.1 | - | 21, 22 | 3257 | ||
| AK077199.1 | - | 6, 7 | 769 | |||
| AK129334.1 | - | 16, 17 | 2607 | |||
| BC072637.1 | - | 20, 21 | 2953 | 30354433 | ||
| CF748039.1 | - | 4, 5 | 423 | |||

Eif2ak4
Genomic Map