siRNA Details
| Gene Symbol: Tlx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019916.2 | NP_064300.2 | 1 | 193 | ||
| GenBank mRNA | AJ223801.1 | - | 1 | 64 | ||
| AK141870.1 | - | 1 | 194 | |||
| BC141308.1 | - | 1 | 123 | 9055920 | ||
| BC145631.1 | - | 1 | 123 | 9054199 | ||

Tlx3
Genomic Map