siRNA Details
| Gene Symbol: Nrk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013724.2 | NP_038752.2 | 6 | 793 | ||
| GenBank mRNA | AB020741.1 | - | 6 | 718 | ||
| AB035267.1 | - | 6 | 471 | |||
| AK041377.1 | - | 6 | 793 | |||
| BC068311.1 | - | 6 | 768 | |||
| BC139029.1 | - | 6 | 560 | 8862051 | ||
| BC139031.1 | - | 6 | 560 | 8862053 | ||

Nrk
Genomic Map