siRNA Details
| Gene Symbol: Mixl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013729.3 | NP_038757.1 | 2 | 670 | ||
| GenBank mRNA | AB221547.1 | - | 2 | 652 | ||
| AF135063.1 | - | 2 | 648 | |||
| AF154573.1 | - | 2 | 670 | |||
| AF201959.1 | - | 2 | 613 | |||
| BC099876.1 | - | 2 | 670 | 40040047 | ||
| BC099877.1 | - | 2 | 670 | 40040048 | ||

Mixl1
Genomic Map