siRNA Details
| Gene Symbol: Sgk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291152.1 | NP_001278081.1 | 6 | 526 | ||
| NM_001291154.1 | NP_001278083.1 | 7 | 1245 | |||
| NM_013731.3 | NP_038759.1 | 7 | 644 | |||
| GenBank mRNA | AF169033.1 | - | 7 | 579 | ||
| AK050009.1 | - | 7 | 1226 | |||
| AK136559.1 | - | 7 | 596 | |||
| BC026549.1 | - | 5 | 271 | 4973908 | ||

Sgk2
Genomic Map