siRNA Details
| Gene Symbol: Smok2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013741.1 | NP_038769.1 | 2 | 1428 | ||
| GenBank mRNA | AJ245453.1 | - | 2 | 1428 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smok2a| Gene Symbol: Smok2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013741.1 | NP_038769.1 | 2 | 1428 | ||
| GenBank mRNA | AJ245453.1 | - | 2 | 1428 | ||