siRNA Details
| Gene Symbol: Zfp354b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013744.3 | NP_038772.1 | 5 | 693 | ||
| GenBank mRNA | AF184112.1 | - | 4 | 562 | ||
| AK148756.1 | - | 5 | 695 | |||
| BC107400.1 | - | 4 | 530 | 40058571 | ||
| BC107401.1 | - | 4 | 530 | 40058574 | ||

Zfp354b
Genomic Map