siRNA Details
| Gene Symbol: Clnk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013748.3 | NP_038776.3 | 17 | 1060 | ||
| GenBank mRNA | AB021220.1 | - | 17 | 1180 | ||
| AF187819.1 | - | 17 | 1060 | |||
| AK082826.1 | - | 13 | 798 | |||
| BC125637.1 | - | 17 | 1057 | 40130152 | ||
| BC125639.1 | - | 17 | 1057 | 40130154 | ||

Clnk
Genomic Map