siRNA Details
| Gene Symbol: Gyg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013755.3 | NP_038783.1 | 4 | 541 | ||
| GenBank mRNA | AF114031.1 | - | 4 | 476 | ||
| AK159576.1 | - | 4 | 522 | |||
| BC029903.1 | - | 4 | 502 | 4487769 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gyg| Gene Symbol: Gyg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013755.3 | NP_038783.1 | 4 | 541 | ||
| GenBank mRNA | AF114031.1 | - | 4 | 476 | ||
| AK159576.1 | - | 4 | 522 | |||
| BC029903.1 | - | 4 | 502 | 4487769 | ||