siRNA Details
| Gene Symbol: Gyg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013755.3 | NP_038783.1 | 3 | 312 | ||
| GenBank mRNA | AA553085.1 | - | 3 | 311 | ||
| AF114031.1 | - | 3 | 247 | |||
| AK159576.1 | - | 3 | 293 | |||
| AK194923.1 | - | 1 | 30 | |||
| AK201075.1 | - | 1 | 30 | |||
| AK208954.1 | - | 1 | 30 | |||
| AK212191.1 | - | 1 | 30 | |||
| AK214082.1 | - | 1 | 30 | |||
| BC029903.1 | - | 3 | 273 | 4487769 | ||

Gyg
Genomic Map