siRNA Details
| Gene Symbol: Csnk1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289898.1 | NP_001276827.1 | 5 | 487 | ||
| NM_001289899.1 | NP_001276828.1 | 5 | 959 | |||
| NM_013767.6 | NP_038795.3 | 5 | 491 | |||
| GenBank mRNA | AB028241.1 | - | 5 | 437 | ||
| AB028736.2 | - | 5 | 437 | |||
| AK158709.1 | - | 5 | 960 | |||
| BC026127.1 | - | 5 | 440 | 4010696 | ||

Csnk1e
Genomic Map