siRNA Details
| Gene Symbol: Akr1c13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013778.2 | NP_038806.2 | 5, 6 | 618 | ||
| GenBank mRNA | AB027125.1 | - | 5, 6 | 584 | ||
| AB097219.1 | - | 5, 6 | 584 | |||
| AK008949.1 | - | 5, 6 | 619 | |||
| AK220165.1 | - | 5, 6 | 611 | |||
| BC021937.1 | - | 5, 6 | 617 | 5100366 | ||
| Gene Symbol: Akr1c12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013777.2 | NP_038805.2 | 5, 6 | 621 | ||
| GenBank mRNA | AB027237.1 | - | 5, 6 | 591 | ||
| AF177041.1 | - | 5, 6 | 612 | |||
| AK007716.1 | - | 5, 6 | 619 | |||
| BC012643.1 | - | 5, 6 | 595 | 4205715 | ||
| BC063780.1 | - | 5, 6 | 598 | 4219625 | ||

Akr1c13
Genomic Map