siRNA Details
| Gene Symbol: Magel2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013779.2 | NP_038807.4 | 1 | 4035 | ||
| GenBank mRNA | AJ243608.1 | - | 1 | 1550 | ||
| AK082944.1 | - | 1 | 4008 | |||
| AK086725.1 | - | 1 | 4030 | |||
| BC054763.1 | - | 1 | 1327 | |||
| BC062913.1 | - | 3 | 3540 | |||
| BC071222.1 | - | 1 | 1813 | |||

Magel2
Genomic Map