siRNA Details
| Gene Symbol: Ip6k1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013785.2 | NP_038813.2 | 2, 3 | 532 | ||
| GenBank mRNA | AF177144.1 | - | 1, 2 | 232 | ||
| AK088872.1 | - | 2, 3 | 549 | |||
| AK166143.1 | - | 2, 3 | 536 | |||
| AK171310.1 | - | 2, 3 | 537 | |||
| AK172921.1 | - | 2, 3 | 591 | |||
| AK180416.1 | - | 1, 2 | 49 | |||
| AK184546.1 | - | 1, 2 | 49 | |||
| AK202332.1 | - | 1, 2 | 73 | |||
| AK204727.1 | - | 1, 2 | 73 | |||
| BC053030.1 | - | 2, 3 | 538 | |||
| BC059006.1 | - | 2, 3 | 532 | 6827203 | ||

Ip6k1
Genomic Map