siRNA Details
| Gene Symbol: Skp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285980.1 | NP_001272909.1 | 9 | 1167 | ||
| NM_013787.3 | NP_038815.1 | 9 | 1237 | |||
| GenBank mRNA | AF083215.1 | - | 9 | 1100 | ||
| AK006221.1 | - | 9 | 1167 | |||
| AK032979.1 | - | 9 | 1148 | |||
| AK044212.1 | - | 9 | 1138 | |||
| AK166034.1 | - | 9 | 1138 | |||
| AK167677.1 | - | 9 | 1135 | |||
| BC092236.1 | - | 9 | 1111 | 30610417 | ||
| BC098371.1 | - | 9 | 1220 | |||
| CK782109.1 | - | 2 | 64 | |||

Skp2
Genomic Map