siRNA Details
| Gene Symbol: Abca3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039581.2 | NP_001034670.1 | 14 | 2367 | ||
| NM_013855.3 | NP_038883.2 | 15 | 2440 | |||
| GenBank mRNA | AK089794.1 | - | 15 | 2381 | ||
| AK154698.1 | - | 15 | 2348 | |||
| AK165458.1 | - | 15 | 2376 | |||
| AK166355.1 | - | 14 | 2347 | |||
| AY083616.1 | - | 15 | 2335 | |||
| BC042663.1 | - | 9 | 1382 | |||
| BC079617.1 | - | 12 | 1931 | 30362309 | ||

Abca3
Genomic Map