siRNA Details
| Gene Symbol: Naglu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013792.2 | NP_038820.2 | 3 | 590 | ||
| GenBank mRNA | AK143546.1 | - | 3 | 591 | ||
| AK158045.1 | - | 1 | 1808 | |||
| AK158774.1 | - | 2 | 1386 | |||
| AK164125.1 | - | 3 | 569 | |||
| AK170758.1 | - | 3 | 573 | |||
| BC055733.1 | - | 3 | 564 | 6411530 | ||
| U85247.1 | - | 3 | 559 | |||

Naglu
Genomic Map