siRNA Details
| Gene Symbol: KCNK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017424.2 | NP_001017424.1 | 2 | 477 | ||
| NM_001017425.2 | NP_001017425.2 | 2 | 278 | |||
| NM_014217.3 | NP_055032.1 | 2 | 154 | |||
| XM_011509522.2 | XP_011507824.1 | 2 | 1680 | |||
| XM_011509523.1 | XP_011507825.1 | 2 | 82 | |||
| XM_011509524.2 | XP_011507826.1 | 2 | 129 | |||
| XM_017001248.1 | XP_016856737.1 | 3 | 865 | |||
| XM_017001249.1 | XP_016856738.1 | 3 | 816 | |||
| GenBank mRNA | AF129399.1 | - | 1 | 64 | ||
| AF171068.1 | - | 2 | 78 | |||
| AK123409.1 | - | 2 | 166 | |||
| AK291483.1 | - | 2 | 258 | |||
| AK294780.1 | - | 2 | 260 | |||
| AK315249.1 | - | 3 | 244 | |||
| AU280191.1 | - | 2 | 220 | |||
| AY552980.2 | - | 2 | 164 | |||
| BC069462.1 | - | 2 | 78 | 7262344 | ||
| BC101693.1 | - | 2 | 123 | 8069199 | ||
| BC101695.1 | - | 2 | 123 | 8069201 | ||
| BC143586.1 | - | 2 | 124 | 9052097 | ||
| CD625291.1 | - | 2 | 471 | |||
| CD625297.1 | - | 2 | 471 | |||
| CD625301.1 | - | 2 | 92 | |||
| DA475072.1 | - | 2 | 478 | |||
| DR001358.1 | - | 2 | 287 | |||
| EF165334.1 | - | 2 | 211 | |||
| EF165335.1 | - | 2 | 176 | |||
| KF182338.1 | - | 2 | 113 | |||
| KF182339.1 | - | 2 | 87 | |||

KCNK2
Genomic Map