siRNA Details
| Gene Symbol: A230006K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK045521.1 | - | 1199 | |||
| DQ656357.1 | - | 2389 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

A230006K03Rik| Gene Symbol: A230006K03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK045521.1 | - | 1199 | |||
| DQ656357.1 | - | 2389 | ||||