siRNA Details
| Gene Symbol: Amot | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153319.3 | NP_695231.3 | 10 | 2534 | ||
| GenBank mRNA | AF461135.1 | - | 10 | 1935 | ||
| AK129277.1 | - | 5 | 774 | |||
| U80888.1 | - | 1 | 106 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amot| Gene Symbol: Amot | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153319.3 | NP_695231.3 | 10 | 2534 | ||
| GenBank mRNA | AF461135.1 | - | 10 | 1935 | ||
| AK129277.1 | - | 5 | 774 | |||
| U80888.1 | - | 1 | 106 | |||