siRNA Details
| Gene Symbol: Nrep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109988.1 | NP_001103458.1 | 5 | 917 | ||
| NM_001109989.2 | NP_001103459.1 | 5 | 840 | |||
| NM_001267717.1 | NP_001254646.1 | 5 | 829 | |||
| NM_053078.4 | NP_444308.1 | 4 | 955 | |||
| GenBank mRNA | AK020629.1 | - | 5 | 776 | ||
| AK048361.1 | - | 4 | 755 | |||
| AK081502.1 | - | 4 | 756 | |||
| AK133769.1 | - | 3 | 2049 | |||
| BC054762.1 | - | 4 | 737 | 6414312 | ||
| BU057200.1 | - | 4 | 743 | |||
| D45203.1 | - | 1 | 352 | |||
| X70398.1 | - | 4 | 752 | |||

Nrep
Genomic Map