siRNA Details
| Gene Symbol: Commd8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178599.4 | NP_848714.1 | 3 | 293 | ||
| GenBank mRNA | AK009275.1 | - | 3 | 282 | ||
| AK012653.1 | - | 3 | 279 | |||
| AK013015.1 | - | 4 | 364 | |||
| AK013371.1 | - | 3 | 266 | |||
| AK077934.1 | - | 3 | 293 | |||
| AK151560.1 | - | 3 | 276 | |||
| AK158078.1 | - | 4 | 376 | |||
| BC051517.1 | - | 1 | 53 | |||
| BC098183.1 | - | 3 | 269 | 5150466 | ||

Commd8
Genomic Map