siRNA Details
| Gene Symbol: Tango2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138583.2 | NP_613049.2 | 3 | 160 | ||
| GenBank mRNA | AK089239.1 | - | 3 | 142 | ||
| AK133588.1 | - | 3 | 143 | |||
| AK168595.1 | - | 2 | 139 | |||
| BC005445.1 | - | 3 | 221 | 3496749 | ||
| X74504.1 | - | 2 | 133 | |||

Tango2
Genomic Map