siRNA Details
| Gene Symbol: Fam21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026585.3 | NP_080861.2 | 10 | 1003 | ||
| GenBank mRNA | AK038318.1 | - | 10 | 981 | ||
| AK122320.1 | - | 9 | 1009 | |||
| AK145716.1 | - | 10 | 1004 | |||
| AK161181.1 | - | 10 | 952 | |||
| AK163765.1 | - | 9 | 991 | |||
| AK168388.1 | - | 10 | 990 | |||
| BC032177.1 | - | 8 | 1230 | |||
| BC040393.1 | - | 10 | 927 | |||
| BC049979.1 | - | 10 | 979 | |||
| BC056942.1 | - | 10 | 930 | 6827177 | ||

Fam21
Genomic Map