siRNA Details
| Gene Symbol: Ubfd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138589.2 | NP_613055.2 | 2 | 551 | ||
| GenBank mRNA | AK011862.1 | - | 2 | 307 | ||
| AK017851.1 | - | 2 | 328 | |||
| AK033428.1 | - | 2 | 543 | |||
| AK077395.1 | - | 2 | 574 | |||
| AK135258.1 | - | 2 | 309 | |||
| AK140891.1 | - | 2 | 310 | |||
| BC011313.1 | - | 2 | 554 | 3499738 | ||

Ubfd1
Genomic Map