siRNA Details
| Gene Symbol: Zcchc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138590.4 | NP_613056.2 | 7, 8 | 1229 | ||
| GenBank mRNA | AK013175.1 | - | 1, 2 | 713 | ||
| AK050430.1 | - | 1, 2 | 1921 | |||
| BC050200.1 | - | 6, 7 | 1104 | |||
| BC054405.1 | - | 1, 2 | 432 | |||

Zcchc7
Genomic Map