siRNA Details
| Gene Symbol: Mrpl50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178603.4 | NP_848718.1 | 2 | 439 | ||
| GenBank mRNA | AK002646.1 | - | 2 | 427 | ||
| AK002686.1 | - | 2 | 441 | |||
| AK007405.1 | - | 2 | 436 | |||
| AK031229.1 | - | 2 | 440 | |||
| AK049421.1 | - | 2 | 429 | |||
| AK075787.1 | - | 2 | 421 | |||
| BC021318.1 | - | 2 | 450 | 5035369 | ||

Mrpl50
Genomic Map