siRNA Details
| Gene Symbol: Gfm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138591.2 | NP_613057.2 | 15 | 1980 | ||
| GenBank mRNA | AF315511.1 | - | 15 | 1947 | ||
| AK199743.1 | - | 1 | 105 | |||
| BC013093.1 | - | 15 | 1983 | 3584730 | ||
| BC031772.1 | - | 15 | 1947 | 4239980 | ||

Gfm1
Genomic Map