siRNA Details
| Gene Symbol: Usp39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138592.4 | NP_613058.1 | 4, 5 | 585 | ||
| GenBank mRNA | AK010443.1 | - | 4, 5 | 595 | ||
| AK018595.1 | - | 4, 5 | 586 | |||
| AK033105.1 | - | 4, 5 | 582 | |||
| AK045062.1 | - | 4, 5 | 582 | |||
| AK088354.1 | - | 4, 5 | 597 | |||
| AK165625.1 | - | 4, 5 | 566 | |||
| AK167668.1 | - | 4, 5 | 573 | |||
| AK168003.1 | - | 4, 5 | 592 | |||
| BC026983.1 | - | 4, 5 | 560 | 5365862 | ||

Usp39
Genomic Map