siRNA Details
| Gene Symbol: Usp39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138592.4 | NP_613058.1 | 2, 3 | 355 | ||
| GenBank mRNA | AK010443.1 | - | 2, 3 | 365 | ||
| AK018595.1 | - | 2, 3 | 356 | |||
| AK033105.1 | - | 2, 3 | 352 | |||
| AK045062.1 | - | 2, 3 | 352 | |||
| AK088354.1 | - | 2, 3 | 367 | |||
| AK165625.1 | - | 2, 3 | 336 | |||
| AK167668.1 | - | 2, 3 | 343 | |||
| AK168003.1 | - | 2, 3 | 362 | |||
| AK201881.1 | - | 2, 3 | 122 | |||
| BC026983.1 | - | 2, 3 | 330 | 5365862 | ||

Usp39
Genomic Map