siRNA Details
| Gene Symbol: Larp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138593.2 | NP_613059.2 | 4 | 465 | ||
| GenBank mRNA | AK012772.1 | - | 4 | 455 | ||
| AK015041.1 | - | 4 | 440 | |||
| AK082815.1 | - | 4 | 464 | |||
| AK162147.1 | - | 4 | 382 | |||
| BC020127.1 | - | 3 | 316 | |||
| BC023165.1 | - | 4 | 419 | |||

Larp7
Genomic Map