siRNA Details
| Gene Symbol: D6Wsu163e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138594.3 | NP_613060.1 | 10 | 1270 | ||
| GenBank mRNA | AK019434.1 | - | 2 | 160 | ||
| AK037095.1 | - | 10 | 1275 | |||
| AK045541.1 | - | 10 | 1271 | |||
| AK046707.1 | - | 10 | 1258 | |||
| AK137201.1 | - | 10 | 1261 | |||
| BC016429.1 | - | 9 | 1218 | 4486831 | ||

D6Wsu163e
Genomic Map