siRNA Details
| Gene Symbol: Fam104a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138598.5 | NP_613064.2 | 3 | 868 | ||
| GenBank mRNA | AK009763.2 | - | 3 | 749 | ||
| AK087540.1 | - | 3 | 779 | |||
| AK145119.1 | - | 3 | 689 | |||
| AK158680.1 | - | 3 | 866 | |||
| AK158872.1 | - | 3 | 915 | |||
| AK159228.1 | - | 3 | 769 | |||
| AK159659.1 | - | 3 | 767 | |||
| BC056429.1 | - | 3 | 610 | |||
| BC060965.1 | - | 3 | 645 | 6705115 | ||

Fam104a
Genomic Map