siRNA Details
| Gene Symbol: Vps25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284411.1 | NP_001271340.1 | 3 | 304 | ||
| NM_001284412.1 | NP_001271341.1 | 3 | 280 | |||
| NM_001284414.1 | NP_001271343.1 | 3 | 280 | |||
| NM_026776.4 | NP_081052.2 | 3 | 280 | |||
| GenBank mRNA | AK002500.1 | - | 3 | 242 | ||
| AK003715.2 | - | 3 | 252 | |||
| AK003877.1 | - | 3 | 238 | |||
| AK082933.1 | - | 3 | 259 | |||
| AK145811.1 | - | 3 | 251 | |||
| AK150822.1 | - | 3 | 262 | |||
| AK162230.1 | - | 3 | 253 | |||
| AK166008.1 | - | 3 | 259 | |||
| AK190530.1 | - | 1 | 34 | |||
| AK214794.1 | - | 1 | 34 | |||
| BC029181.1 | - | 3 | 244 | 4240500 | ||

Vps25
Genomic Map