siRNA Details
| Gene Symbol: Rtcb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145422.4 | NP_663397.1 | 9 | 1123 | ||
| GenBank mRNA | AK145891.1 | - | 9 | 1120 | ||
| AK145916.1 | - | 9 | 1120 | |||
| AK149885.1 | - | 9 | 1078 | |||
| AK150376.1 | - | 9 | 1082 | |||
| AK150470.1 | - | 9 | 1082 | |||
| AK151042.1 | - | 9 | 1116 | |||
| AK151182.1 | - | 9 | 1082 | |||
| AK151932.1 | - | 9 | 1082 | |||
| AK151988.1 | - | 9 | 1116 | |||
| AK152144.1 | - | 9 | 1082 | |||
| AK153434.1 | - | 9 | 1082 | |||
| AK167643.1 | - | 9 | 1109 | |||
| AK168038.1 | - | 9 | 1117 | |||
| AK172175.1 | - | 9 | 1117 | |||
| AK191745.1 | - | 2 | 77 | |||
| AY424364.1 | - | 9 | 1116 | |||
| BC003288.1 | - | 9 | 1125 | 3583081 | ||

Rtcb
Genomic Map