siRNA Details
| Gene Symbol: Rtcb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145422.4 | NP_663397.1 | 6 | 664 | ||
| GenBank mRNA | AK145891.1 | - | 6 | 661 | ||
| AK145916.1 | - | 6 | 661 | |||
| AK149885.1 | - | 6 | 619 | |||
| AK150376.1 | - | 6 | 623 | |||
| AK150470.1 | - | 6 | 623 | |||
| AK151042.1 | - | 6 | 657 | |||
| AK151182.1 | - | 6 | 623 | |||
| AK151932.1 | - | 6 | 623 | |||
| AK151988.1 | - | 6 | 658 | |||
| AK152144.1 | - | 6 | 623 | |||
| AK153434.1 | - | 6 | 623 | |||
| AK167643.1 | - | 6 | 650 | |||
| AK168038.1 | - | 6 | 658 | |||
| AK172175.1 | - | 6 | 658 | |||
| AK199892.1 | - | 2 | 225 | |||
| AK206521.1 | - | 2 | 206 | |||
| AK208576.1 | - | 2 | 207 | |||
| AK218642.1 | - | 2 | 206 | |||
| AY424364.1 | - | 6 | 657 | |||
| BC003288.1 | - | 6 | 666 | 3583081 | ||

Rtcb
Genomic Map