siRNA Details
| Gene Symbol: Trim36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170855.1 | NP_001164326.1 | 1, 2 | 316 | ||
| NM_178872.4 | NP_849203.2 | 1, 2 | 317 | |||
| GenBank mRNA | AB103063.1 | - | 1, 2 | 111 | ||
| AY251389.1 | - | 1, 2 | 74 | |||
| BC058106.1 | - | 1, 2 | 235 | 6836197 | ||
| CJ046908.1 | - | 1, 2 | 315 | |||
| CJ167273.1 | - | 1, 2 | 318 | |||

Trim36
Genomic Map