siRNA Details
| Gene Symbol: Tinf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145705.3 | NP_663751.2 | 5 | 776 | ||
| GenBank mRNA | AF214013.1 | - | 5 | 767 | ||
| AF518764.1 | - | 5 | 741 | |||
| AK016456.1 | - | 5 | 709 | |||
| AK028952.1 | - | 5 | 747 | |||
| AK090324.1 | - | 5 | 743 | |||
| AK144592.1 | - | 5 | 705 | |||
| AK169480.1 | - | 3 | 291 | |||
| AK205058.1 | - | 1 | 35 | |||
| BC017142.1 | - | 2 | 1051 | |||
| BC030347.1 | - | 5 | 702 | 5003804 | ||

Tinf2
Genomic Map