siRNA Details
| Gene Symbol: Reep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204915.1 | NP_001191844.1 | 2 | 230 | ||
| NM_178606.5 | NP_848721.1 | 2 | 230 | |||
| GenBank mRNA | AK005026.1 | - | 2 | 178 | ||
| AK008058.1 | - | 2 | 149 | |||
| AK012403.1 | - | 2 | 174 | |||
| AK047728.1 | - | 2 | 98 | |||
| AK079649.1 | - | 2 | 100 | |||
| AK154559.1 | - | 2 | 88 | |||
| AY562231.1 | - | 2 | 63 | |||
| BC004607.1 | - | 2 | 139 | 3585133 | ||
| CJ163373.1 | - | 2 | 232 | |||

Reep3
Genomic Map