siRNA Details
| Gene Symbol: Reep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204915.1 | NP_001191844.1 | 6, 7 | 737 | ||
| NM_178606.5 | NP_848721.1 | 6, 7 | 737 | |||
| GenBank mRNA | AK005026.1 | - | 6, 7 | 683 | ||
| AK008058.1 | - | 6, 7 | 656 | |||
| AK012403.1 | - | 6, 7 | 681 | |||
| AK028868.1 | - | 6, 7 | 694 | |||
| AK047728.1 | - | 6, 7 | 605 | |||
| AK079649.1 | - | 6, 7 | 607 | |||
| AK154559.1 | - | 6, 7 | 595 | |||
| AY562231.1 | - | 6, 7 | 570 | |||
| BC004607.1 | - | 6, 7 | 646 | 3585133 | ||

Reep3
Genomic Map