siRNA Details
| Gene Symbol: Reep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204915.1 | NP_001191844.1 | 3 | 300 | ||
| NM_178606.5 | NP_848721.1 | 3 | 300 | |||
| GenBank mRNA | AK005026.1 | - | 3 | 248 | ||
| AK008058.1 | - | 3 | 219 | |||
| AK012403.1 | - | 3 | 244 | |||
| AK047728.1 | - | 3 | 168 | |||
| AK079649.1 | - | 3 | 170 | |||
| AK154559.1 | - | 3 | 158 | |||
| AY562231.1 | - | 3 | 133 | |||
| BC004607.1 | - | 3 | 209 | 3585133 | ||
| CJ163373.1 | - | 3 | 302 | |||

Reep3
Genomic Map