siRNA Details
| Gene Symbol: Trpm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138301.2 | NP_612174.2 | 29 | 4257 | ||
| GenBank mRNA | AB166747.1 | - | 29 | 4239 | ||
| AJ344343.1 | - | 27 | 4388 | |||
| AJ878415.1 | - | 27 | 3940 | |||
| AK034099.1 | - | 6 | 528 | |||
| AK036731.1 | - | 11 | 2536 | |||
| AK134735.1 | - | 17 | 2398 | |||
| AK136905.1 | - | 12 | 2311 | |||
| BC141391.1 | - | 27 | 3970 | 9056004 | ||

Trpm2
Genomic Map