siRNA Details
| Gene Symbol: D10Jhu81e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138601.2 | NP_613067.1 | 5 | 475 | ||
| GenBank mRNA | AK003866.1 | - | 5 | 476 | ||
| BC013475.1 | - | 5 | 468 | 4237589 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

D10Jhu81e| Gene Symbol: D10Jhu81e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138601.2 | NP_613067.1 | 5 | 475 | ||
| GenBank mRNA | AK003866.1 | - | 5 | 476 | ||
| BC013475.1 | - | 5 | 468 | 4237589 | ||