siRNA Details
| Gene Symbol: Limd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013860.2 | NP_038888.2 | 3 | 2051 | ||
| GenBank mRNA | AJ132409.1 | - | 3 | 2040 | ||
| AK004806.1 | - | 3 | 1355 | |||
| AK047198.1 | - | 3 | 2045 | |||
| AK083970.1 | - | 3 | 2013 | |||
| AK180850.1 | - | 2 | 142 | |||
| AK190733.1 | - | 2 | 143 | |||
| AK193947.1 | - | 2 | 142 | |||
| AK194248.1 | - | 2 | 141 | |||
| AK197187.1 | - | 2 | 143 | |||
| AK197198.1 | - | 2 | 142 | |||
| AK207336.1 | - | 2 | 142 | |||
| AK210590.1 | - | 2 | 142 | |||
| AK217088.1 | - | 2 | 142 | |||
| BC056449.1 | - | 3 | 1926 | 5703026 | ||

Limd1
Genomic Map