siRNA Details
| Gene Symbol: Zfp385a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013866.2 | NP_038894.2 | 3 | 225 | ||
| GenBank mRNA | AF118566.1 | - | 3 | 223 | ||
| BC017644.1 | - | 3 | 282 | 4504518 | ||
| CK619761.1 | - | 3 | 223 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp385a| Gene Symbol: Zfp385a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013866.2 | NP_038894.2 | 3 | 225 | ||
| GenBank mRNA | AF118566.1 | - | 3 | 223 | ||
| BC017644.1 | - | 3 | 282 | 4504518 | ||
| CK619761.1 | - | 3 | 223 | |||